Assay Details
| Gene Symbol: Gm9159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033398.1 | 1 | 1544 | 74 | ||
| GenBank mRNA | AK084556.1 | - | 1 | 1545 | 74 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AJ421479.1 | - | Chr. - - [-] on Build | 74 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | polycomb group ring finger 6 pseudogene | Gm9785 | ENSMUSG00000043661 | -19.7kb | Anti-sense | Chr. X - 103525319 - 103525847 on NCBI Build GRCm38 |
| Downstream |
Ftx transcript, Xist regulator (non-protein coding) |
Ftx |
B230206F22Rik, BB219131, NCRNA00182, Thcytx |
13.2kb | Anti-sense | Chr. X - 103560910 - 103623754 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm9159
Genomic Map