Assay Details
| Gene Symbol: AU019990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033468.1 | 5 - 6 | 616 | 63 | ||
| GenBank mRNA | AK166556.1 | - | 5 - 6 | 619 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glycerophosphocholine phosphodiesterase GDE1 homolog (S. cerevisiae) | Gpcpd1 |
2310004G06Rik, 2310032D16Rik, AU015213, Gde5, Prei4, mKIAA1434 |
-69.7kb | Anti-sense | Chr. 2 - 132529082 - 132578282 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1110034G24 gene | 1110034G24Rik | 33.8kb | Sense | Chr. 2 - 132686875 - 132751055 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK166556.1 transcripts.
Set Membership: |
|

AU019990
Genomic Map