Assay Details
| Gene Symbol: Gm10768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033472.1 | 1 | 455 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATP-binding cassette, sub-family C (CFTR/MRP), member 2 |
Abcc2 |
AI173996, Abc30, Cmoat, Mrp2, cMRP |
-471.0 | Sense | Chr. 19 - 43782308 - 43838332 on NCBI Build GRCm38 |
| Downstream | dynamin binding protein | Dnmbp |
2410003L07Rik, 2410003M15Rik, TUBA |
6.0kb | Anti-sense | Chr. 19 - 43846815 - 43940318 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm10768
Genomic Map