Assay Details
| Gene Symbol: Gm13939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033473.1 | 3 - 4 | 395 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lin-7 homolog C (C. elegans) | Lin7c |
9130007B12Rik, AI303698, AU019331, AW125731, D2Ertd520e, LIN-7-C, LIN-7C, MALS-3, Veli3 |
-5.6kb | Anti-sense | Chr. 2 - 109890878 - 109900937 on NCBI Build GRCm38 |
| Downstream | leucine-rich repeat-containing G protein-coupled receptor 4 | Lgr4 |
9130225G07, A330106J01Rik, A930009A08Rik, Gpr48 |
9.6kb | Anti-sense | Chr. 2 - 109917647 - 110014257 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0300003839.rn
Set Membership: |
|

Gm13939
Genomic Map