Assay Details
| Gene Symbol: Gm10823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033475.1 | 4 - 5 | 315 | 102 | ||
| GenBank mRNA | AK136620.1 | - | 4 - 5 | 316 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 50 | Ccdc50 | C3orf6 | -463.0kb | Sense | Chr. 16 - 27388765 - 27452218 on NCBI Build GRCm38 |
| Downstream | fibroblast growth factor 12 | Fgf12 |
AV114868, B230343J05Rik, FGF-12, FHF-1, Fhf1 |
231.7kb | Anti-sense | Chr. 16 - 28157783 - 28753243 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK136620.1 transcripts.
Set Membership: |
|

Gm10823
Genomic Map