Assay Details
| Gene Symbol: Gm10510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033511.1 | 1 - 2 | 94 | 89 | ||
| GenBank mRNA | CJ072309.1 | - | 1 - 2 | 94 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 5091 | Gm5091 | C330048F19 | -110.9kb | Sense | Chr. 17 - 15231585 - 15240783 on NCBI Build GRCm38 |
| Downstream | delta-like 1 (Drosophila) | Dll1 | Delta1 | 12.9kb | Sense | Chr. 17 - 15367354 - 15376113 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10510
Genomic Map