Assay Details
| Gene Symbol: Gm11529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033524.1 | 1 - 2 | 460 | 76 | ||
| GenBank mRNA | AK138406.1 | - | 1 - 2 | 461 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox B1 | Hoxb1 | Hox-2.9 | -95.3kb | Anti-sense | Chr. 11 - 96365746 - 96368254 on NCBI Build GRCm38 |
| Downstream | src family associated phosphoprotein 1 | Skap1 |
1700091G21Rik, Scap1, Skap-55 |
29.0 | Anti-sense | Chr. 11 - 96464576 - 96759622 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK138406.1 transcripts.
Set Membership: |
|

Gm11529
Genomic Map