Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10474512 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10474513 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10474511 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0200012298.mm.1 |
NR_033534.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012298.mm.1 |
TC0200001652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012293.mm.1 |
NR_033534.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012293.mm.1 |
TC0200001652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012297.mm.1 |
NR_033534.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012297.mm.1 |
TC0200001652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012294.mm.1 |
NR_033534.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012294.mm.1 |
TC0200001652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012292.mm.1 |
NR_033534.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012292.mm.1 |
TC0200001652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012296.mm.1 |
NR_033534.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012296.mm.1 |
TC0200001652.mm.1 |
MTA TC |