Assay Details
| Gene Symbol: Gm10516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033536.1 | 1 - 2 | 165 | 59 | ||
| GenBank mRNA | AK142593.1 | - | 1 - 2 | 166 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TNF receptor-associated factor 5 | Traf5 | -44.3kb | Anti-sense | Chr. 1 - 192052688 - 192092599 on NCBI Build GRCm38 | |
| Downstream |
potassium voltage-gated channel, subfamily H (eag-related), member 1 |
Kcnh1 |
EAG1, Kv10.1, M-eag |
45.8kb | Sense | Chr. 1 - 192190362 - 192510159 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10516
Genomic Map