Assay Details
| Gene Symbol: Gm10516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033536.1 | 2 - 3 | 265 | 86 | ||
| GenBank mRNA | AK142593.1 | - | 2 - 3 | 266 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | REST corepressor 3 | Rcor3 |
4921514E24Rik, C730034D20Rik, E130101E15Rik |
-6.4kb | Anti-sense | Chr. 1 - 192098546 - 192138067 on NCBI Build GRCm38 |
| Downstream |
potassium voltage-gated channel, subfamily H (eag-related), member 1 |
Kcnh1 |
EAG1, Kv10.1, M-eag |
44.9kb | Sense | Chr. 1 - 192190362 - 192510159 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10516
Genomic Map