Assay Details
| Gene Symbol: Gm14812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033544.2 | 1 - 2 | 76 | 91 | ||
| GenBank mRNA | AK135730.1 | - | 1 - 2 | 77 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 14809 | Gm14809 | OTTMUSG00000018062 | -22.8kb | Sense | Chr. X - 99407579 - 99414219 on NCBI Build GRCm38 |
| Downstream |
praja ring finger 1, E3 ubiquitin protein ligase |
Pja1 | praja-1 | 21.9kb | Sense | Chr. X - 99465734 - 99471268 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK135730.1 transcripts.
Set Membership: |
|

Gm14812
Genomic Map