Assay Details
| Gene Symbol: Gm14812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033544.2 | 6 - 7 | 659 | 57 | ||
| GenBank mRNA | AK135730.1 | - | 6 - 7 | 661 | 57 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 14809 | Gm14809 | OTTMUSG00000018062 | Sense | Chr. X - 99407579 - 99414219 on NCBI Build GRCm38 | |
| Downstream |
praja ring finger 1, E3 ubiquitin protein ligase |
Pja1 | praja-1 | 33.6kb | Sense | Chr. X - 99465734 - 99471268 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK135730.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2129916 | NR_033544.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2129916 | AK135730.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281315 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628845 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 178182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12115 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560791 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193583 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630488 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143376 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589832 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269257 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423395 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 158346 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158114 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2129916 | NR_033544.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2129916 | AK135730.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03724356 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158114 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03724336 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14785078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15180419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14934691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14707048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14902843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15202487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14893164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14813636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14629383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14535671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14593192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14593187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14693966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14887000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14693964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15180426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14720096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14615254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14548535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15142515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15142519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14838174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14535665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14902845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14548530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15153289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14552894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14516860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14725764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14870986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15020794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14775449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15083778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14632441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14819450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15094378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14813638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15083784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14535669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14934695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15207335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14934698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14934694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14899223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14707046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14899216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32555366 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.1 ST Array | 10605917 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00016911.mm.1 | NR_033544.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016911.mm.1 | TC0X00002699.mm.1 | MTA TC |

Gm14812
Genomic Map