Assay Details
| Gene Symbol: Gm15348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033546.1 | 1 - 2 | 98 | 102 | ||
| GenBank mRNA | AK143914.1 | - | 1 - 2 | 99 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
tubulin, gamma complex associated protein 3 |
Tubgcp3 |
GCP3, Spc98p |
-46.4kb | Sense | Chr. 8 - 12614277 - 12672243 on NCBI Build GRCm38 |
| Downstream |
ATPase, class VI, type 11A |
Atp11a |
4930558F19Rik, AU040868, Atpc1h, Ih |
37.9kb | Anti-sense | Chr. 8 - 12757016 - 12868728 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK143914.1 transcripts.
Set Membership: |
|

Gm15348
Genomic Map