Assay Details
| Gene Symbol: Gm12359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033551.1 | 2 - 3 | 481 | 88 | ||
| GenBank mRNA | AK142978.1 | - | 2 - 3 | 482 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
nuclear receptor subfamily 1, group D, member 1 |
Nr1d1 |
A530070C09Rik, R75201 |
-22.9kb | Sense | Chr. 11 - 98767932 - 98775377 on NCBI Build GRCm38 |
| Downstream | cancer susceptibility candidate 3 | Casc3 |
Btz, Mln51 |
7.8kb | Anti-sense | Chr. 11 - 98809808 - 98833807 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm12359
Genomic Map