Assay Details
| Gene Symbol: Gm8221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033577.1 | 1 | 1847 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | apolipoprotein L 10B | Apol10b | 9130218O11Rik | -20.4kb | Anti-sense | Chr. 15 - 77584106 - 77598542 on NCBI Build GRCm38 |
| Downstream | apolipoprotein L 11b | Apol11b | A330102K04Rik | 6.6kb | Anti-sense | Chr. 15 - 77633951 - 77643286 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm8221
Genomic Map