Assay Details
| Gene Symbol: Gm11426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033582.1 | 1 - 2 | 129 | 95 | ||
| GenBank mRNA | AY512941.1 | - | 1 - 2 | 71 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 132E | Tmem132e | Gm644 | -185.7kb | Sense | Chr. 11 - 82387446 - 82447618 on NCBI Build GRCm38 |
| Downstream |
chaperonin containing Tcp1, subunit 6b (zeta) |
Cct6b |
CCT-zeta-2, CCTzeta-2, Cctz-2, TCP-1-zeta-2 |
85.5kb | Anti-sense | Chr. 11 - 82719248 - 82764321 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY512941.1 transcripts.
Set Membership: |
|

Gm11426
Genomic Map