Assay Details
| Gene Symbol: A930017M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033609.2 | 1 | 2645 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 2310069G16 gene | 2310069G16Rik | -75.6kb | Sense | Chr. 15 - 44787763 - 44805829 on NCBI Build GRCm38 | |
| Downstream |
potassium channel, subfamily V, member 1 |
Kcnv1 |
2700023A03Rik, vibe |
221.5kb | Anti-sense | Chr. 15 - 45106237 - 45116488 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A930017M01Rik
Genomic Map