Assay Details
| Gene Symbol: Cyp2c53-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033614.1 | 1 - 2 | 207 | 73 | ||
| GenBank mRNA | AY227735.1 | - | 1 - 2 | 207 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
cytochrome P450, family 2, subfamily c, polypeptide 66 |
Cyp2c66 | 2010301M18Rik | -42.5kb | Sense | Chr. 19 - 39113898 - 39186756 on NCBI Build GRCm38 |
| Downstream |
cytochrome P450, family 2, subfamily c, polypeptide 29 |
Cyp2c29 |
AHOH, AHOHase, AI159681, AI196010, Ah-2, Ahh-1, Cyp2c, P450-2C |
45.7kb | Sense | Chr. 19 - 39287044 - 39330713 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp2c53-ps
Genomic Map