Assay Details
| Gene Symbol: Gm8709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033633.1 | 1 | 449 | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | l(3)mbt-like 3 (Drosophila) | L3mbtl3 |
AI481284, MBT-1 |
-297.5kb | Sense | Chr. 10 - 26274470 - 26375989 on NCBI Build GRCm38 |
| Downstream | Rho GTPase activating protein 18 | Arhgap18 |
4833419J07Rik, AI314906, AI843501 |
20.7kb | Anti-sense | Chr. 10 - 26695312 - 26918648 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm8709
Genomic Map