Assay Details
| Gene Symbol: 1700044K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033785.1 | 1 - 2 | 68 | 89 | ||
| GenBank mRNA | AK006696.1 | - | 1 - 2 | 69 | 89 | |
| AK018859.1 | - | 1 - 2 | 69 | 89 | ||
| AK018880.1 | - | 1 - 2 | 69 | 89 | ||
| BY706767.1 | - | 1 - 2 | 69 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA G630055G22 gene | G630055G22Rik | -1.4mb | Sense | Chr. 18 - 47922841 - 48107980 on NCBI Build GRCm38 | |
| Downstream | DTW domain containing 2 | Dtwd2 |
1190002H09Rik, 8030470C17Rik, AI414673, BB115449 |
171.7kb | Anti-sense | Chr. 18 - 49696145 - 49755667 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1800000483.rn
Set Membership: |
|

1700044K03Rik
Genomic Map