Assay Details
| Gene Symbol: Mir1941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035462.1 | 1 | 44 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 35853 |
Gm35853 | Sense | Chr. 15 - 101358708 - 101405857 on NCBI Build GRCm38 | ||
| Downstream | keratin 7 | Krt7 |
D15Wsu77e, K7, Krt2-7 |
43.0kb | Sense | Chr. 15 - 101412403 - 101427806 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir1941
Genomic Map