Assay Details
| Gene Symbol: Mir1945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035466.1 | 1 | 40 | 62 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G1 to S phase transition 1 | Gspt1 |
AI314175, AI326449, AV307676, C79774, G1st, Gst-1 |
-43.0 | Sense | Chr. 16 - 11216240 - 11254325 on NCBI Build GRCm38 |
| Downstream |
tumor necrosis factor receptor superfamily, member 17 |
Tnfrsf17 |
BCM, BCMA, Tnfrsf13, Tnfrsf13a |
58.7kb | Anti-sense | Chr. 16 - 11313174 - 11320074 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir1945
Genomic Map