Assay Details
| Gene Symbol: Mir1947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035469.1 | 1 | 39 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L35A | Rpl35a |
2810431L15Rik, Rpl35 |
-45.2kb | Sense | Chr. 16 - 33056453 - 33060189 on NCBI Build GRCm38 |
| Downstream | oxysterol binding protein-like 11 | Osbpl11 |
9430097N02Rik, AI132306, AU019795, ORP-11 |
64.3kb | Sense | Chr. 16 - 33169763 - 33243312 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir1947
Genomic Map