Assay Details
| Gene Symbol: Mir1983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035500.1 | 1 | 65 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 40830 |
Gm40830 | Anti-sense | Chr. 13 - 21896116 - 21908096 on NCBI Build GRCm38 | ||
| Downstream | zinc finger protein 184 (Kruppel-like) | Zfp184 | 4930500C15Rik | 47.9kb | Anti-sense | Chr. 13 - 21944940 - 21960495 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Mir1983
Genomic Map