Assay Details
| Gene Symbol: Mir599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035527.1 | 1 | 44 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC048602 | BC048602 | -332.3kb | Sense | Chr. 15 - 35307010 - 35328535 on NCBI Build GRCm38 | |
| Downstream | microRNA 875 | Mir875 |
Mirn875, mir-875, mmu-mir-875 |
52.0 | Sense | Chr. 15 - 35660970 - 35661047 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir599
Genomic Map