Assay Details
| Gene Symbol: Gm5084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036449.1 | 2 - 3 | 1502 | 65 | ||
| GenBank mRNA | AK037190.1 | - | 2 - 3 | 1503 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | growth arrest specific 1 | Gas1 |
AW554192, Gas-1 |
-31.1kb | Anti-sense | Chr. 13 - 60174405 - 60177535 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA A530001N23 gene | A530001N23Rik | 270.8kb | Anti-sense | Chr. 13 - 60483921 - 60508840 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037190.1 transcripts.
Set Membership: |
|

Gm5084
Genomic Map