Assay Details
| Gene Symbol: Gm4814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036451.1 | 2 - 3 | 225 | 100 | ||
| GenBank mRNA | AK050630.1 | - | 2 - 3 | 226 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 31630 |
Gm31630 | -164.9kb | Sense | Chr. 13 - 92883010 - 92908723 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA A630019I02 gene | A630019I02Rik | 20.1kb | Sense | Chr. 13 - 93094833 - 93098968 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050630.1 transcripts.
Set Membership: |
|

Gm4814
Genomic Map