Assay Details
| Gene Symbol: Mir3075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037236.1 | 1 | 43 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Zmiz1 opposite strand 1 | Zmiz1os1 |
C130026E14, D930049A15Rik, Zmiz1as1 |
-75.1kb | Anti-sense | Chr. 14 - 25242913 - 25459339 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4931406H21 gene | 4931406H21Rik | 52.3kb | Sense | Chr. 14 - 25586804 - 25590661 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3075
Genomic Map