Assay Details
| Gene Symbol: Mir3093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037276.1 | 1 | 36 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 3764 | Gm3764 |
100042271, Gm3759 |
Sense | Chr. 3 - 88205769 - 88229959 on NCBI Build GRCm38 | |
| Downstream | microRNA 9-1 | Mir9-1 |
Mirn9, Mirn9-1, mir-9-1, mmu-mir-9-1 |
341.0 | Sense | Chr. 3 - 88215598 - 88215686 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3093
Genomic Map