Assay Details
| Gene Symbol: Mir3095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037278.1 | 1 | 46 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
muscle, skeletal, receptor tyrosine kinase |
Musk |
Mdk4, Mlk, Nsk1, Nsk2, Nsk3 |
-63.2kb | Anti-sense | Chr. 4 - 58285640 - 58377848 on NCBI Build GRCm38 |
| Downstream | olfactory receptor 267 | Olfr267 | MOR262-1 | 342.0kb | Sense | Chr. 4 - 58783130 - 58788108 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3095
Genomic Map