Assay Details
| Gene Symbol: Mir3105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037292.1 | 1 | 40 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6404 | Mir6404 |
Gm28014, mmu-mir-6404 |
-76.0kb | Sense | Chr. 7 - 143933115 - 143933208 on NCBI Build GRCm38 |
| Downstream | cortactin | Cttn |
1110020L01Rik, Ems1 |
426.4kb | Anti-sense | Chr. 7 - 144435723 - 144471004 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3105
Genomic Map