Assay Details
| Gene Symbol: Atp5j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AA259963.1 | - | 2 - 3 | 209 | 103 | |
| AK078484.1 | - | 2 - 3 | 370 | 103 | ||
| BC010766.1 | - | 3 - 4 | 505 | 4219715 | 103 | |
| BE851144.1 | - | 3 - 4 | 247 | 103 | ||
| BY079469.1 | - | 2 - 3 | 284 | 103 | ||
| U77128.1 | - | 1 - 2 | 168 | 103 | ||
| W75852.1 | - | 2 - 3 | 235 | 103 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30925872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30925871 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30925875 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30925869 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963772 | NR_002690.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720090 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 793110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57854 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154471 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963772 | NR_002690.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P100866 | AK078484.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P100866 | BC010766.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P100866 | U77128.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P100866 | NM_016755.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30925878 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08667315 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154471 | - | - |
| Whole Mouse Genome G4122A | A_51_P100866 | AK078484.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100866 | BC010766.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100866 | U77128.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100866 | NM_016755.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119351 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115224 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154471 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12923791 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12524196 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30925871 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30925872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30925875 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30925869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30925908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12521612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12538183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12959087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12943097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13022696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30925895 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10440487 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440484 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440485 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416143_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10440483 | - | - |
| Mouse Genome 430 2.0 Array | 1416143_at | - | - |
| Mouse Genome 430 2.0 Array | 1416143_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11957_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11957_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600015204.mm.1 | NM_016755.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015204.mm.1 | TC1600001964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600015219.mm.1 | NM_016755.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015219.mm.1 | TC1600001964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600015220.mm.1 | NM_016755.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015220.mm.1 | TC1600001964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600015221.mm.1 | NM_016755.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015221.mm.1 | TC1600001964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600015214.mm.1 | NM_016755.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015214.mm.1 | TC1600001964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600015199.mm.1 | NM_016755.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015199.mm.1 | TC1600001964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600015203.mm.1 | NM_016755.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015203.mm.1 | TC1600001964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600015213.mm.1 | NM_016755.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015213.mm.1 | TC1600001964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600015212.mm.1 | NM_016755.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015212.mm.1 | TC1600001964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600015197.mm.1 | NM_016755.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015197.mm.1 | TC1600001964.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416143_PM_at | - | - |
| Mouse Expression Set 430 | 1416143_at | - | - |
| Murine Genome U74v2 Set | 98128_at | - | - |
| Murine Genome U74v2 Set | 98128_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416143_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416143_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4210100 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6900288 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004210100 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900288 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2632890 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231207 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2632890 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1212748 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231207 | - | - |
| mouse-6 v1.1 (Illumina) | scl0000102.1_16-S | - | - |
| mouse-6 v1.1 (Illumina) | scl48278.6.1_15-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4210100 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900288 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0000102.1_16-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl48278.6.1_15-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4210100 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900288 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2611261 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2632890 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2962632 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2611261 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2632890 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2962632 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0000102.1_16-S | - | - |

Atp5j
Genomic Map