Assay Details
| Gene Symbol: Gm6042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002872.1 | 1 - 2 | 109 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 5430421N21 gene | 5430421N21Rik | Krt83 | -12.3kb | Anti-sense | Chr. 15 - 101485130 - 101491512 on NCBI Build GRCm38 |
| Downstream | keratin 84 | Krt84 |
AV235125, HRb-1, Krt2-16, Krt2-3 |
10.4kb | Anti-sense | Chr. 15 - 101516663 - 101532959 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm6042
Genomic Map