Assay Details
| Gene Symbol: Gm8234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_004433.3 | 1 | 1943 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glutamate rich 6 | Erich6 |
4932431H17Rik, Fam194a |
Anti-sense | Chr. 3 - 58616300 - 58665413 on NCBI Build GRCm38 | |
| Downstream | seven in absentia 2 | Siah2 |
AA415433, Sinh2 |
20.9kb | Anti-sense | Chr. 3 - 58674949 - 58692388 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm8234
Genomic Map