Assay Details
Gene Symbol: Mir3110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037297.1 | 1 | 42 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 255A | Tmem255a |
4933417N17, 6430550H21Rik, Fam70a |
Anti-sense | Chr. X - 38196573 - 38252489 on NCBI Build GRCm38 | |
Downstream |
ATPase, (Na+)/K+ transporting, beta 4 polypeptide |
Atp1b4 | 1110020B02Rik | 105.6kb | Sense | Chr. X - 38316122 - 38336784 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
