Assay Details
| Gene Symbol: 1600020E01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037960.1 | 1 - 2 | 107 | 96 | ||
| GenBank mRNA | AA162444.1 | - | 1 - 2 | 105 | 96 | |
| AI644794.1 | - | 1 - 2 | 103 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | poly(rC) binding protein 1 | Pcbp1 |
WBP17, [a]CP-1, alphaCP-1 |
-1.2kb | Anti-sense | Chr. 6 - 86524494 - 86526171 on NCBI Build GRCm38 |
| Downstream |
aspartic peptidase, retroviral-like 1 |
Asprv1 |
2300003P22Rik, AA986851, SASP, SASPase, Taps |
100.2kb | Sense | Chr. 6 - 86628174 - 86629710 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1600020E01Rik
Genomic Map