Assay Details
| Gene Symbol: 1600020E01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037960.1 | 2 - 3 | 192 | 83 | ||
| NR_037961.1 | 2 - 3 | 219 | 83 | |||
| GenBank mRNA | AA162444.1 | - | 2 - 3 | 190 | 83 | |
| AI644794.1 | - | 2 - 3 | 188 | 83 | ||
| AK005503.1 | - | 2 - 3 | 220 | 83 | ||
| BC029726.1 | - | 1709 | 83 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | poly(rC) binding protein 1 | Pcbp1 |
WBP17, [a]CP-1, alphaCP-1 |
-1.7kb | Anti-sense | Chr. 6 - 86524494 - 86526171 on NCBI Build GRCm38 |
| Downstream |
aspartic peptidase, retroviral-like 1 |
Asprv1 |
2300003P22Rik, AA986851, SASP, SASPase, Taps |
95.9kb | Sense | Chr. 6 - 86628174 - 86629710 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1600020E01Rik
Genomic Map