| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2356840 |
BC029726.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
78753 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
188303 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
337944 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
581270 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
135510 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
425020 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
379710 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577785 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
353364 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
69345 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
87445 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
AK033082.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
AK135613.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
AK138854.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
AK143464.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
AK146676.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
AK156563.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
BC024537.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
BC089523.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
BC099952.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001472029.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001472053.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001472087.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001472530.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001472797.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001472832.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001473776.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001475495.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001475694.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001475762.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001475779.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001476309.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001477605.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001478015.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001478221.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001478340.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001478377.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001478477.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001478679.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001479521.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001479562.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001479624.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001479755.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001480229.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001480397.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001480743.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001480950.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001481117.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_001481156.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900403 |
XM_618937.3 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
620168 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
941378 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
525223 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
379244 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
272123 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
384586 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
658648 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
969517 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
113330 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
60142 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01375670 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05969158 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01375690 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01375661 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05969181 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P617171 |
AA162444.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P617171 |
AI448276.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P617171 |
NR_037961.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P617171 |
AI644794.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P617171 |
AK005503.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P128256 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01375690 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05969158 |
- |
- |