Assay Details
| Gene Symbol: Mir3070b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037231.1 | 1 | 51 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3070a | Mir3070a |
mir-3070-1, mmu-mir-3070-1, mmu-mir-3070a |
-561.0 | Sense | Chr. 12 - 109587943 - 109588031 on NCBI Build GRCm38 |
| Downstream | retrotransposon-like 1 | Rtl1 |
6430411K18Rik, Mar, Mart1, Mor1, Peg11 |
1.5kb | Anti-sense | Chr. 12 - 109590169 - 109595403 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3070b
Genomic Map