Assay Details
| Gene Symbol: Airn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027773.1 | 3 | 451 | 94 | ||
| GenBank mRNA | AK013242.1 | - | 3 | 391 | 94 | |
| BY718621.1 | - | 3 | 452 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MAS1 oncogene | Mas1 |
Mas-1, Mgra |
Anti-sense | Chr. 17 - 12838841 - 12868145 on NCBI Build GRCm38 | |
| Downstream |
MAS-related GPR, member H |
Mrgprh |
Gpr90, Mgrb, MrgH |
16.2kb | Sense | Chr. 17 - 12876034 - 12877842 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Airn
Genomic Map