Assay Details
| Gene Symbol: B230208H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038027.1 | 1 | 51 | 88 | ||
| NR_038028.1 | 1 | 51 | 88 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | utrophin | Utrn |
AA589569, DRP, Dmdl |
-52.9kb | Sense | Chr. 10 - 12382187 - 12869685 on NCBI Build GRCm38 |
| Downstream | syntaxin 11 | Stx11 | 5830405C08Rik | 16.9kb | Sense | Chr. 10 - 12939983 - 12964500 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

B230208H11Rik
Genomic Map