Assay Details
| Gene Symbol: Eid2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177427.1 | NP_001170898.1 | 1 | 351 | 77 | |
| GenBank mRNA | AK044722.1 | - | 1 | 320 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eid2b
Genomic Map