Assay Details
| Gene Symbol: Gm1110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281475.1 | NP_001268404.1 | 16 - 17 | 1613 | 89 | |
| GenBank mRNA | AK136683.1 | - | 7 - 8 | 1039 | 89 | |
| DQ664197.1 | - | 16 - 17 | 1622 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gm1110
Genomic Map