Assay Details
| Gene Symbol: Gm1110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281475.1 | NP_001268404.1 | 4 - 5 | 442 | 101 | |
| GenBank mRNA | DQ664197.1 | - | 4 - 5 | 442 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gm1110
Genomic Map