Assay Details
| Gene Symbol: Gps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177874.1 | NP_001171345.1 | 5 - 6 | 643 | 83 | |
| NM_145370.2 | NP_663345.2 | 5 - 6 | 783 | 83 | ||
| GenBank mRNA | AK088490.1 | - | 5 - 6 | 640 | 83 | |
| BC003350.1 | - | 5 - 6 | 783 | 3481979 | 83 | |
| BC103782.1 | - | 5 - 6 | 610 | 6486599 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gps1
Genomic Map