Assay Details
| Gene Symbol: Junb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008416.3 | NP_032442.1 | 1 | 1050 | 91 | |
| GenBank mRNA | AK044253.1 | - | 1021 | 91 | ||
| AK088643.1 | - | 1 | 1021 | 91 | ||
| AK141592.1 | - | 1 | 1021 | 91 | ||
| AK144871.1 | - | 1 | 1028 | 91 | ||
| AK145550.1 | - | 1 | 1020 | 91 | ||
| AK149794.1 | - | 1 | 1023 | 91 | ||
| AK150111.1 | - | 1 | 1021 | 91 | ||
| AK150247.1 | - | 1 | 1021 | 91 | ||
| AK150286.1 | - | 1 | 1020 | 91 | ||
| AK155043.1 | - | 1 | 1018 | 91 | ||
| AK159083.1 | - | 1 | 1020 | 91 | ||
| AK159140.1 | - | 1 | 1019 | 91 | ||
| AK159520.1 | - | 1 | 1020 | 91 | ||
| AK159661.1 | - | 1 | 1022 | 91 | ||
| AK160066.1 | - | 1 | 1021 | 91 | ||
| BC003790.1 | - | 1 | 999 | 3592704 | 91 | |
| BC092302.1 | - | 1 | 997 | 6399302 | 91 | |
| J03236.1 | - | 1 | 1050 | 91 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Junb
Genomic Map