Assay Details
| Gene Symbol: Hus1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153072.2 | NP_694712.1 | 1 | 249 | 95 | |
| GenBank mRNA | AF508546.1 | - | 1 | 162 | 95 | |
| AK133107.1 | - | 1 | 250 | 95 | ||
| BC125607.1 | - | 1 | 242 | 40130122 | 95 | |
| BC125609.1 | - | 1 | 242 | 40130124 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hus1b
Genomic Map