Assay Details
| Gene Symbol: Rae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175112.5 | NP_780321.1 | 3 - 4 | 463 | 67 | |
| GenBank mRNA | AK014325.1 | - | 3 - 4 | 420 | 67 | |
| AK017746.1 | - | 3 - 4 | 384 | 67 | ||
| AK035278.1 | - | 3 - 4 | 409 | 67 | ||
| AK079383.1 | - | 3 - 4 | 444 | 67 | ||
| AK146058.1 | - | 3 - 4 | 428 | 67 | ||
| AK156681.1 | - | 3 - 4 | 428 | 67 | ||
| BC059051.1 | - | 3 - 4 | 385 | 6818771 | 67 | |
| BC060072.1 | - | 3 - 4 | 330 | 5718934 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rae1
Genomic Map