Assay Details
| Gene Symbol: Rae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175112.5 | NP_780321.1 | 6 - 7 | 722 | 94 | |
| GenBank mRNA | AK035278.1 | - | 6 - 7 | 668 | 94 | |
| AK079383.1 | - | 6 - 7 | 703 | 94 | ||
| AK146058.1 | - | 6 - 7 | 687 | 94 | ||
| AK156681.1 | - | 6 - 7 | 687 | 94 | ||
| AK209630.1 | - | 3 - 4 | 185 | 94 | ||
| BC059051.1 | - | 6 - 7 | 644 | 6818771 | 94 | |
| BC060072.1 | - | 6 - 7 | 589 | 5718934 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rae1
Genomic Map