Assay Details
| Gene Symbol: Itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010584.3 | NP_034714.1 | 6 - 7 | 772 | 100 | |
| GenBank mRNA | AB016496.1 | - | 6 - 7 | 771 | 100 | |
| AK007447.1 | - | 6 - 7 | 772 | 100 | ||
| AY157363.1 | - | 6 - 7 | 761 | 100 | ||
| AY157364.1 | - | 6 - 7 | 756 | 100 | ||
| BC150795.1 | - | 6 - 7 | 714 | 9087706 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itln1
Genomic Map